7-Chloro-2-ethyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

Supplier: Matrix Scientific

080500-500MG 080500-1G
102669-892EA 387.69 USD
102669-892 102669-894
7-Chloro-2-ethyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
7-Chloro-2-ethyl-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine

Hazard: Irritant

Formula: C₈H₉ClN₄
MW: 196.64 g/mol
MDL Number: MFCD13811403
CAS Number: 28565-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR