Methyl {(4E)-5-oxo-1-phenyl-4-[1-(1H-pyrazol-3-ylamino)ethylidene]-4,5-dihydro-1H-pyrazol-3-yl}acetate

Supplier: Matrix Scientific

080493-500MG 080493-1G
102669-870EA 193.85 USD
102669-870 102669-872
Methyl {(4E)-5-oxo-1-phenyl-4-[1-(1H-pyrazol-3-ylamino)ethylidene]-4,5-dihydro-1H-pyrazol-3-yl}acetate
Methyl {(4E)-5-oxo-1-phenyl-4-[1-(1H-pyrazol-3-ylamino)ethylidene]-4,5-dihydro-1H-pyrazol-3-yl}acetate

Hazard: Irritant

Formula: C₁₇H₁₇N₅O₃
MDL Number: MFCD22374891
CAS Number: 1160226-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR