2-Chloro-4,5,6,7-tetrahydro-1H-benzimidazole

Supplier: Matrix Scientific

080488-500MG 080488-1G
102669-854EA 438.46 USD
102669-854 102669-856
2-Chloro-4,5,6,7-tetrahydro-1H-benzimidazole
2-Chloro-4,5,6,7-tetrahydro-1H-benzimidazole

Hazard: Irritant

Formula: C₇H₉ClN₂
MDL Number: MFCD13811162
CAS Number: 1049127-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR