Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-(2-furylmethyl)-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Supplier: Matrix Scientific
080487-500MG
080487-1G
102669-850EA
193.85
USD
102669-850
102669-852
Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-(2-furylmethyl)-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-(2-furylmethyl)-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Hazard: Irritant
Formula:
C₁₉H₁₉N₃O₄ |
MDL Number:
MFCD22374889 CAS Number: 1374510-90-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...