Methyl-6-[2-(3,4-dimethoxyphenyl)ethyl]-2-[(E)-2-(dimethylamino)-vinyl]-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Supplier: Matrix Scientific

080486-500MG 080486-1G
102669-846EA 193.85 USD
102669-846 102669-848
Methyl-6-[2-(3,4-dimethoxyphenyl)ethyl]-2-[(E)-2-(dimethylamino)-vinyl]-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-6-[2-(3,4-dimethoxyphenyl)ethyl]-2-[(E)-2-(dimethylamino)-vinyl]-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₂₄H₂₇N₃O₅
MDL Number: MFCD22374888
CAS Number: 1374510-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR