Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-(4-methoxyphenyl)-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Supplier: Matrix Scientific

080485-500MG 080485-1G
102669-842EA 193.85 USD
102669-842 102669-844
Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-(4-methoxyphenyl)-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-(4-methoxyphenyl)-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₂₁H₂₁N₃O₄
MDL Number: MFCD22374887
CAS Number: 1374510-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR