Methyl-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-6-phenyl-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Supplier: Matrix Scientific
080484-500MG
080484-1G
080484-5G
102669-836EA
193.85
USD
102669-836
102669-838
102669-840
Methyl-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-6-phenyl-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-6-phenyl-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Hazard: Irritant
Formula:
C₂₀H₁₉N₃O₃ |
MDL Number:
MFCD22374886 CAS Number: 1374510-87-8 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...