Methyl-6-(3,5-dichlorophenyl)-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Supplier: Matrix Scientific

080483-500MG 080483-1G
102669-832EA 193.85 USD
102669-832 102669-834
Methyl-6-(3,5-dichlorophenyl)-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-6-(3,5-dichlorophenyl)-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₂₀H₁₇Cl₂N₃O₃
MDL Number: MFCD22374885
CAS Number: 1374510-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR