Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-isopropyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Supplier: Matrix Scientific

080482-500MG 080482-1G
102669-828EA 193.85 USD
102669-828 102669-830
Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-isopropyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-2-[(E)-2-(dimethylamino)vinyl]-6-isopropyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₁N₃O₃
MDL Number: MFCD22374884
CAS Number: 1374510-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR