7-Amino-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile

Supplier: Matrix Scientific

080471-500MG 080471-1G
102669-796EA 387.69 USD
102669-796 102669-798
7-Amino-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile
7-Amino-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carbonitrile

Hazard: Irritant

Formula: C₇H₃F₃N₆
MDL Number: MFCD07657834
CAS Number: 890091-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR