Methyl (3-oxopyrazolidin-4-yl)acetate
Supplier: Matrix Scientific
080469-500MG
080469-1G
102669-788EA
516.92
USD
102669-788
102669-790
Methyl (3-oxopyrazolidin-4-yl)acetate
Methyl (3-oxopyrazolidin-4-yl)acetate
Hazard: Irritant
Formula:
C₆H₁₀N₂O₃ |
MDL Number:
MFCD13817362 CAS Number: 1114595-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...