1-[2-(4-Ethylpiperazin-1-yl)-4-methylpyrimidin-5-yl]ethanone

Supplier: Matrix Scientific

080468-500MG 080468-1G
102669-784EA 258.46 USD
102669-784 102669-786
1-[2-(4-Ethylpiperazin-1-yl)-4-methylpyrimidin-5-yl]ethanone
1-[2-(4-Ethylpiperazin-1-yl)-4-methylpyrimidin-5-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₂₀N₄O
MDL Number: MFCD08131973
CAS Number: 903447-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR