1-[4-Methyl-2-(4-methylpiperidin-1-yl)pyrimidin-5-yl]ethanone

Supplier: Matrix Scientific

080465-500MG 080465-1G 080465-5G
102669-770EA 258.46 USD
102669-770 102669-772 102669-774
1-[4-Methyl-2-(4-methylpiperidin-1-yl)pyrimidin-5-yl]ethanone
1-[4-Methyl-2-(4-methylpiperidin-1-yl)pyrimidin-5-yl]ethanone

Hazard: Irritant

Formula: C₁₃H₁₉N₃O
MDL Number: MFCD08131964
CAS Number: 903447-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR