2-[(2,4-Dichlorophenyl)amino]-5,6-dimethylpyrimidin-4(3H)-one

Supplier: Matrix Scientific

080454-500MG 080454-1G
102669-738EA 193.85 USD
102669-738 102669-740
2-[(2,4-Dichlorophenyl)amino]-5,6-dimethylpyrimidin-4(3H)-one
2-[(2,4-Dichlorophenyl)amino]-5,6-dimethylpyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₁₂H₁₁Cl₂N₃O
MW: 284.15 g/mol
MDL Number: MFCD22374874
CAS Number: 1374509-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR