[1,2,4]Triazolo[5,1-c][1,2,4]benzotriazin-6-ol
Supplier: Matrix Scientific
080453-500MG
080453-1G
102669-734EA
387.69
USD
102669-734
102669-736
[1,2,4]Triazolo[5,1-c][1,2,4]benzotriazin-6-ol
[1,2,4]Triazolo[5,1-c][1,2,4]benzotriazin-6-ol
Hazard: Irritant
Formula:
C₈H₅N₅O |
MDL Number:
MFCD22374873 CAS Number: 1374509-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...