Methyl-4-[(E)-2-(dimethylamino)vinyl]pyrazolo[5,1-c][1,2,4]triazine-3-carboxylate

Supplier: Matrix Scientific

080452-500MG 080452-1G
102669-730EA 258.46 USD
102669-730 102669-732
Methyl-4-[(E)-2-(dimethylamino)vinyl]pyrazolo[5,1-c][1,2,4]triazine-3-carboxylate
Methyl-4-[(E)-2-(dimethylamino)vinyl]pyrazolo[5,1-c][1,2,4]triazine-3-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₃N₅O₂
MDL Number: MFCD22374872
CAS Number: 1374510-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR