1-{7-[(E)-2-(Dimethylamino)vinyl]pyrazolo[1,5-a]pyrimidin-6-yl}ethanone

Supplier: Matrix Scientific

080449-500MG 080449-1G
102669-720EA 387.69 USD
102669-720 102669-722
1-{7-[(E)-2-(Dimethylamino)vinyl]pyrazolo[1,5-a]pyrimidin-6-yl}ethanone
1-{7-[(E)-2-(Dimethylamino)vinyl]pyrazolo[1,5-a]pyrimidin-6-yl}ethanone

Hazard: Irritant

Formula: C₁₂H₁₄N₄O
MDL Number: MFCD22374869
CAS Number: 130506-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR