1-[7-[(E)-2-(Dimethylamino)vinyl]-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-6-yl]ethanone

Supplier: Matrix Scientific

080448-500MG 080448-1G 080448-5G
102669-714EA 193.85 USD
102669-714 102669-716 102669-718
1-[7-[(E)-2-(Dimethylamino)vinyl]-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-6-yl]ethanone
1-[7-[(E)-2-(Dimethylamino)vinyl]-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidin-6-yl]ethanone

Hazard: Irritant

Formula: C₁₂H₁₂F₃N₅O
MDL Number: MFCD22374868
CAS Number: 1374510-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR