Ethyl-4-[(E)-2-(dimethylamino)vinyl]pyrimido[1,2-a]benzimidazole-3-carboxylate

Supplier: Matrix Scientific

080447-500MG 080447-1G
102669-710EA 193.85 USD
102669-710 102669-712
Ethyl-4-[(E)-2-(dimethylamino)vinyl]pyrimido[1,2-a]benzimidazole-3-carboxylate
Ethyl-4-[(E)-2-(dimethylamino)vinyl]pyrimido[1,2-a]benzimidazole-3-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₈N₄O₂
MDL Number: MFCD22374867
CAS Number: 1374510-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR