Ethyl-7-[(E)-2-(dimethylamino)vinyl]-2-(methoxymethyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Supplier: Matrix Scientific

080446-500MG 080446-1G 080446-5G
102669-704EA 193.85 USD
102669-704 102669-706 102669-708
Ethyl-7-[(E)-2-(dimethylamino)vinyl]-2-(methoxymethyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl-7-[(E)-2-(dimethylamino)vinyl]-2-(methoxymethyl)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₉N₅O₃
MDL Number: MFCD22374866
CAS Number: 1374510-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR