Dimethyl-7-[(E)-2-(dimethylamino)vinyl]-2-methylpyrazolo[1,5-a]pyrimidine-3,6-dicarboxylate

Supplier: Matrix Scientific

080443-500MG 080443-1G 080443-5G
102669-690EA 193.85 USD
102669-690 102669-692 102669-694
Dimethyl-7-[(E)-2-(dimethylamino)vinyl]-2-methylpyrazolo[1,5-a]pyrimidine-3,6-dicarboxylate
Dimethyl-7-[(E)-2-(dimethylamino)vinyl]-2-methylpyrazolo[1,5-a]pyrimidine-3,6-dicarboxylate

Hazard: Irritant

Formula: C₁₅H₁₈N₄O₄
MDL Number: MFCD22374863
CAS Number: 1374510-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR