Ethyl-7-[(E)-2-(dimethylamino)vinyl]-2-(ethylthio)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Supplier: Matrix Scientific

080442-500MG 080442-1G 080442-5G
102669-684EA 193.85 USD
102669-684 102669-686 102669-688
Ethyl-7-[(E)-2-(dimethylamino)vinyl]-2-(ethylthio)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl-7-[(E)-2-(dimethylamino)vinyl]-2-(ethylthio)[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₉N₅O₂S
MDL Number: MFCD22374862
CAS Number: 1374510-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR