Methyl-3-[2-(4-ethylpiperazin-1-yl)-4-methylpyrimidin-5-yl]-3-oxopropanoate
Supplier: Matrix Scientific
080441-500MG
080441-1G
080441-5G
102669-678EA
193.85
USD
102669-678
102669-680
102669-682
Methyl-3-[2-(4-ethylpiperazin-1-yl)-4-methylpyrimidin-5-yl]-3-oxopropanoate
Methyl-3-[2-(4-ethylpiperazin-1-yl)-4-methylpyrimidin-5-yl]-3-oxopropanoate
Hazard: Irritant
Formula:
C₁₅H₂₂N₄O₃ |
MDL Number:
MFCD22374861 CAS Number: 1374509-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...