7-[(Dimethylamino)methyl]-2,2,4-trimethyl-1,2-dihydroquinolin-6-ol

Supplier: Matrix Scientific

080436-500MG 080436-1G 080436-5G
102669-656EA 258.46 USD
102669-656 102669-658 102669-660
7-[(Dimethylamino)methyl]-2,2,4-trimethyl-1,2-dihydroquinolin-6-ol
7-[(Dimethylamino)methyl]-2,2,4-trimethyl-1,2-dihydroquinolin-6-ol

Hazard: Irritant

Formula: C₁₅H₂₂N₂O
MDL Number: MFCD00532547
CAS Number: 300690-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR