[1-(Tetrahydro-2H-pyran-4-yl)cyclopropyl]amine hydrochloride

Supplier: Matrix Scientific

080431-500MG 080431-1G 080431-5G
102669-626EA 523.08 USD
102669-626 102669-628 102669-630
[1-(Tetrahydro-2H-pyran-4-yl)cyclopropyl]amine hydrochloride
[1-(Tetrahydro-2H-pyran-4-yl)cyclopropyl]amine hydrochloride

Hazard: Irritant

Formula: C₈H₁₆ClNO
MDL Number: MFCD22421982
CAS Number: 1417568-40-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR