[1-(Tetrahydrofuran-3-yl)cyclopropyl]amine hydrochloride

Supplier: Matrix Scientific

080430-500MG 080430-1G 080430-5G
102669-620EA 775.38 USD
102669-620 102669-622 102669-624
[1-(Tetrahydrofuran-3-yl)cyclopropyl]amine hydrochloride
[1-(Tetrahydrofuran-3-yl)cyclopropyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO
MDL Number: MFCD22421981
CAS Number: 1417568-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR