[1-Cyclopropyl-2-(3-methylphenoxy)ethyl]amine

Supplier: Matrix Scientific

080423-500MG 080423-1G
102669-586EA 516.92 USD
102669-586 102669-588
[1-Cyclopropyl-2-(3-methylphenoxy)ethyl]amine
[1-Cyclopropyl-2-(3-methylphenoxy)ethyl]amine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD19605918
CAS Number: 1343224-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR