[1-Cyclopropyl-2-(3,4-dimethylphenoxy)ethyl]amine

Supplier: Matrix Scientific

080422-500MG 080422-1G
102669-582EA 516.92 USD
102669-582 102669-584
[1-Cyclopropyl-2-(3,4-dimethylphenoxy)ethyl]amine
[1-Cyclopropyl-2-(3,4-dimethylphenoxy)ethyl]amine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD19605912
CAS Number: 1341558-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR