[1-Cyclopropyl-2-(2-naphthyloxy)ethyl]amine

Supplier: Matrix Scientific

080421-500MG 080421-1G 080421-5G
102669-576EA 387.69 USD
102669-576 102669-578 102669-580
[1-Cyclopropyl-2-(2-naphthyloxy)ethyl]amine
[1-Cyclopropyl-2-(2-naphthyloxy)ethyl]amine

Hazard: Irritant

Formula: C₁₅H₁₇NO
MDL Number: MFCD19546754
CAS Number: 1379811-36-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR