2-(Biphenyl-4-yloxy)-1-cyclopropylethanol

Supplier: Matrix Scientific

080420-500MG 080420-1G 080420-5G
102669-570EA 258.46 USD
102669-570 102669-572 102669-574
2-(Biphenyl-4-yloxy)-1-cyclopropylethanol
2-(Biphenyl-4-yloxy)-1-cyclopropylethanol

Hazard: Irritant

Formula: C₁₇H₁₈O₂
MDL Number: MFCD19391588
CAS Number: 1379811-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR