[1-Cyclopropyl-2-(2-methylphenoxy)ethyl]amine
Supplier: Matrix Scientific
080417-500MG
080417-1G
102669-556EA
516.92
USD
102669-556
102669-558
[1-Cyclopropyl-2-(2-methylphenoxy)ethyl]amine
[1-Cyclopropyl-2-(2-methylphenoxy)ethyl]amine
Hazard: Irritant
Formula:
C₁₂H₁₇NO |
MDL Number:
MFCD19605922 CAS Number: 1342595-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...