[2-(2-Chloro-5-methylphenoxy)-1-cyclopropylethyl]methylamine

Supplier: Matrix Scientific

080415-500MG 080415-1G
102669-548EA 387.69 USD
102669-548 102669-550
[2-(2-Chloro-5-methylphenoxy)-1-cyclopropylethyl]methylamine
[2-(2-Chloro-5-methylphenoxy)-1-cyclopropylethyl]methylamine

Hazard: Irritant

Formula: C₁₃H₁₈ClNO
MDL Number: MFCD22374859
CAS Number: 1379811-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR