[1-Cyclopropyl-2-(3-ethoxyphenoxy)ethyl]methylamine

Supplier: Matrix Scientific

080414-500MG 080414-1G
102669-544EA 387.69 USD
102669-544 102669-546
[1-Cyclopropyl-2-(3-ethoxyphenoxy)ethyl]methylamine
[1-Cyclopropyl-2-(3-ethoxyphenoxy)ethyl]methylamine

Hazard: Irritant

Formula: C₁₄H₂₁NO₂
MDL Number: MFCD19055693
CAS Number: 1379811-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR