[1-Cyclopropyl-2-(3,4-dichlorophenoxy)ethyl]methylamine

Supplier: Matrix Scientific

080413-500MG 080413-1G 080413-5G
102669-538EA 387.69 USD
102669-538 102669-540 102669-542
[1-Cyclopropyl-2-(3,4-dichlorophenoxy)ethyl]methylamine
[1-Cyclopropyl-2-(3,4-dichlorophenoxy)ethyl]methylamine

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂NO
MDL Number: MFCD22374858
CAS Number: 1379811-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR