[1-Cyclopropyl-2-(3,5-dichlorophenoxy)ethyl]methylamine

Supplier: Matrix Scientific

080412-500MG 080412-1G 080412-5G
102669-532EA 387.69 USD
102669-532 102669-534 102669-536
[1-Cyclopropyl-2-(3,5-dichlorophenoxy)ethyl]methylamine
[1-Cyclopropyl-2-(3,5-dichlorophenoxy)ethyl]methylamine

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂NO
MDL Number: MFCD22374857
CAS Number: 1379811-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR