[1-Cyclopropyl-2-(3-fluorophenoxy)ethyl]methylamine

Supplier: Matrix Scientific

080411-500MG 080411-1G
102669-528EA 516.92 USD
102669-528 102669-530
[1-Cyclopropyl-2-(3-fluorophenoxy)ethyl]methylamine
[1-Cyclopropyl-2-(3-fluorophenoxy)ethyl]methylamine

Hazard: Irritant

Formula: C₁₂H₁₆FNO
MDL Number: MFCD19605942
CAS Number: 1341502-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR