[1-Cyclopropyl-2-(2,4-dimethylphenoxy)ethyl]amine

Supplier: Matrix Scientific

080410-500MG 080410-1G 080410-5G
102669-522EA 516.92 USD
102669-522 102669-524 102669-526
[1-Cyclopropyl-2-(2,4-dimethylphenoxy)ethyl]amine
[1-Cyclopropyl-2-(2,4-dimethylphenoxy)ethyl]amine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD19605870
CAS Number: 1340391-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR