[1-Cyclopropyl-2-(4-ethylphenoxy)ethyl]amine

Supplier: Matrix Scientific

080409-500MG 080409-1G
102669-518EA 516.92 USD
102669-518 102669-520
[1-Cyclopropyl-2-(4-ethylphenoxy)ethyl]amine
[1-Cyclopropyl-2-(4-ethylphenoxy)ethyl]amine

Hazard: Irritant

Formula: C₁₃H₁₉NO
MDL Number: MFCD19605893
CAS Number: 1341840-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR