[1-Cyclopropyl-2-(4-methoxyphenoxy)ethyl]amine
Supplier: Matrix Scientific
080408-500MG
080408-1G
102669-514EA
516.92
USD
102669-514
102669-516
[1-Cyclopropyl-2-(4-methoxyphenoxy)ethyl]amine
[1-Cyclopropyl-2-(4-methoxyphenoxy)ethyl]amine
Hazard: Irritant
Formula:
C₁₂H₁₇NO₂ |
MDL Number:
MFCD19605946 CAS Number: 1341809-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...