[2-(4-Chloro-3-methylphenoxy)-1-cyclopropylethyl]amine hydrochloride
Supplier: Matrix Scientific
080406-500MG
080406-1G
080406-5G
102669-504EA
516.92
USD
102669-504
102669-506
102669-508
[2-(4-Chloro-3-methylphenoxy)-1-cyclopropylethyl]amine hydrochloride
[2-(4-Chloro-3-methylphenoxy)-1-cyclopropylethyl]amine hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇Cl₂NO |
MDL Number:
MFCD22421978 CAS Number: 1417566-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...