{2-[(4-Chlorophenyl)thio]-1-cyclopropylethyl}methylamine

Supplier: Matrix Scientific

080404-500MG 080404-1G 080404-5G
102669-494EA 516.92 USD
102669-494 102669-496 102669-498
{2-[(4-Chlorophenyl)thio]-1-cyclopropylethyl}methylamine
{2-[(4-Chlorophenyl)thio]-1-cyclopropylethyl}methylamine

Hazard: Irritant

Formula: C₁₂H₁₆ClNS
MDL Number: MFCD20373427
CAS Number: 1340419-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR