{1-Cyclopropyl-2-[(4-methylphenyl)thio]ethyl}methylamine

Supplier: Matrix Scientific

080403-500MG 080403-1G
102669-490EA 516.92 USD
102669-490 102669-492
{1-Cyclopropyl-2-[(4-methylphenyl)thio]ethyl}methylamine
{1-Cyclopropyl-2-[(4-methylphenyl)thio]ethyl}methylamine

Hazard: Irritant

Formula: C₁₃H₁₉NS
MDL Number: MFCD20373740
CAS Number: 1344302-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR