1-Cyclopropyl-N~2~-ethyl-N~2~-phenylethane-1,2-diamine

Supplier: Matrix Scientific

080402-500MG 080402-1G 080402-5G
102669-484EA 516.92 USD
102669-484 102669-486 102669-488
1-Cyclopropyl-N~2~-ethyl-N~2~-phenylethane-1,2-diamine
1-Cyclopropyl-N~2~-ethyl-N~2~-phenylethane-1,2-diamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD19604692
CAS Number: 1343166-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR