1-Cyclopropyl-N~2~-methyl-N~2~-phenylethane-1,2-diamine

Supplier: Matrix Scientific

080401-500MG 080401-1G 080401-5G
102669-478EA 516.92 USD
102669-478 102669-480 102669-482
1-Cyclopropyl-N~2~-methyl-N~2~-phenylethane-1,2-diamine
1-Cyclopropyl-N~2~-methyl-N~2~-phenylethane-1,2-diamine

Hazard: Irritant

Formula: C₁₂H₁₈N₂
MDL Number: MFCD19604655
CAS Number: 1342365-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR