1-Cyclopropyl-N~2~,N~2~-diethyl-N~1~-methylethane-1,2-diamine
Supplier: Matrix Scientific
080400-500MG
080400-1G
080400-5G
102669-472EA
516.92
USD
102669-472
102669-474
102669-476
1-Cyclopropyl-N~2~,N~2~-diethyl-N~1~-methylethane-1,2-diamine
1-Cyclopropyl-N~2~,N~2~-diethyl-N~1~-methylethane-1,2-diamine
Hazard: Irritant
Formula:
C₁₀H₂₂N₂ MW: 170.3 g/mol |
MDL Number:
MFCD19604644 CAS Number: 1341066-00-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...