(1-Cyclopropyl-2-morpholin-4-ylethyl)amine

Supplier: Matrix Scientific

080399-500MG 080399-1G
102669-468EA 646.15 USD
102669-468 102669-470
(1-Cyclopropyl-2-morpholin-4-ylethyl)amine
(1-Cyclopropyl-2-morpholin-4-ylethyl)amine

Hazard: Irritant

Formula: C₉H₁₈N₂O
MDL Number: MFCD19604639
CAS Number: 1343839-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR