(1-Cyclopropyl-2-piperidin-1-ylethyl)methylamine

Supplier: Matrix Scientific

080397-500MG 080397-1G
102669-458EA 516.92 USD
102669-458 102669-460
(1-Cyclopropyl-2-piperidin-1-ylethyl)methylamine
(1-Cyclopropyl-2-piperidin-1-ylethyl)methylamine

Hazard: Irritant

Formula: C₁₁H₂₂N₂
MDL Number: MFCD19604677
CAS Number: 1342479-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR