[1-Cyclopropyl-2-(propylthio)ethyl]methylamine

Supplier: Matrix Scientific

080395-500MG 080395-1G 080395-5G
102669-448EA 516.92 USD
102669-448 102669-450 102669-452
[1-Cyclopropyl-2-(propylthio)ethyl]methylamine
[1-Cyclopropyl-2-(propylthio)ethyl]methylamine

Hazard: Irritant

Formula: C₉H₁₉NS
MDL Number: MFCD20373323
CAS Number: 1339355-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR