[1-Cyclopropyl-2-(ethylthio)ethyl]methylamine

Supplier: Matrix Scientific

080394-500MG 080394-1G 080394-5G
102669-442EA 516.92 USD
102669-442 102669-444 102669-446
[1-Cyclopropyl-2-(ethylthio)ethyl]methylamine
[1-Cyclopropyl-2-(ethylthio)ethyl]methylamine

Hazard: Irritant

Formula: C₈H₁₇NS
MDL Number: MFCD20373011
CAS Number: 1339109-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR