[2-(1-Methyl-1H-pyrazol-5-yl)ethyl]amine
Supplier: Matrix Scientific
080392-500MG
080392-1G
080392-5G
102669-430EA
646.15
USD
102669-430
102669-432
102669-434
[2-(1-Methyl-1H-pyrazol-5-yl)ethyl]amine
[2-(1-Methyl-1H-pyrazol-5-yl)ethyl]amine
Hazard: Irritant
Formula:
C₆H₁₁N₃ |
MDL Number:
MFCD19620954 CAS Number: 807315-40-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...